missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-4-chloro-3-indolyl beta-D-glucoside, 99%

Product Code. 15497029
Change view
Click to view available options
Quantity:
500 mg
1 g
5 g
Unit Size:
1g
500mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15497029 1 g 1g
15470467 500 mg 500mg
15407039 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15497029 Supplier Thermo Scientific Alfa Aesar Supplier No. J65677.03
 Restricted Item
Estimated Shipment: 15-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Substrate for beta→-D-glucosidase | CAS: 15548-60-4 | C14H15BrClNO6 | 408.63 g/mol

Substrate for beta--D-glucosidase

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15548-60-4
Molecular Formula C14H15BrClNO6
Molecular Weight (g/mol) 408.63
MDL Number MFCD00063690
InChI Key OPIFSICVWOWJMJ-LNNRFACYSA-N
Synonym x-glc, x-glucoside, 5-bromo-4-chloro-3-indolyl-beta-d-glucoside, 5-bromo-4-chloro-3-indolyl-beta-d-glucopyranoside, 5-bromo-4-chloroindol-3-yl-beta-d-glucopyranoside, 5-bromo-4-chloro-3-indolyl b-d-glucopyranoside, 5-bromo-4-chloro-3-indolyl beta-d-glucoside, 2s,3r,4s,5s,6r-2-5-bromo-4-chloro-1h-indol-3-yl oxy-6-hydroxymethyl tetrahydro-2h-pyran-3,4,5-triol, 2s,3r,4s,5s,6r-2-5-bromo-4-chloro-1h-indol-3-yl oxy-6-hydroxymethyl oxane-3,4,5-triol, indoxyl-glucoside
PubChem CID 84982
ChEBI CHEBI:75424
IUPAC Name (2S,3R,4S,5S,6R)-2-[(5-bromo-4-chloro-1H-indol-3-yl)oxy]-6-(hydroxymethyl)oxane-3,4,5-triol
SMILES OC[C@H]1O[C@@H](OC2=CNC3=CC=C(Br)C(Cl)=C23)[C@H](O)[C@@H](O)[C@@H]1O

Specifications

Color White
Quantity 1 g
Beilstein 1552603
Solubility Information Soluble in DMF or DMSO
Formula Weight 408.63
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 5-Bromo-4-chloro-3-indolyl beta-D-glucoside

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.