missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-4-chloro-3-indolyl-beta-D-galactopyranoside (X-GAL), >99%, Ultrapure, Thermo Scientific™

Product Code. 15835048 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
2 g
5 g
Packaging:
Plastic bottle
Unit Size:
100mg
1g
250mg
2g
5g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15835048 5 g Plastic bottle 5g
15825248 100 mg Plastic bottle 100mg
15835248 250 mg Plastic bottle 250mg
15815048 1 g Plastic bottle 1g
15825048 2 g Plastic bottle 2g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 15835048 Supplier Thermo Scientific Supplier No. J10077.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Histochemical substrate for β-Galactosidase and substrate for blue/white colony screening

A histochemical substrate for β-Galactosidase. A substrate for blue/white colony screening. Effective concentration is approximately 50 μg/ml.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7240-90-6
Molecular Formula C14H15BrClNO6
Molecular Weight (g/mol) 408.629
MDL Number MFCD00005666
InChI Key OPIFSICVWOWJMJ-AEOCFKNESA-N
Synonym x-gal, 5-bromo-4-chloro-3-beta-d-galactopyranosyloxy indole, 5-bromo-4-chloro-3-indolyl-beta-d-galactoside, unii-v595og374w, 5-bromo-4-chloro-3-indolyl beta-galactoside, 5-bromo-4-chloroindol-3-yl-beta-d-galactopyranoside, 5-bromo-4-chloro-3-indolyl beta-d-galactoside, 5-bromo-4-chloro-3-indolyl-beta-d-galactopyranoside, 5-bromo-4-chloro-3-indolyl beta-d-galactopyranoside, indoxyl-gal
PubChem CID 65181
ChEBI CHEBI:75055
IUPAC Name (2S,3R,4S,5R,6R)-2-[(5-bromo-4-chloro-1H-indol-3-yl)oxy]-6-(hydroxymethyl)oxane-3,4,5-triol
SMILES C1=CC(=C(C2=C1NC=C2OC3C(C(C(C(O3)CO)O)O)O)Cl)Br

Specifications

Melting Point ∼236°C (decomposition)
Packaging Plastic bottle
Quantity 5 g
Formula Weight 408.63
Percent Purity ≥99%
Grade Ultrapure
Physical Form Crystalline Powder
Chemical Name or Material 5-Bromo-4-chloro-3-indolyl-beta-D-galactopyranoside (X-GAL)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.