missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-4-chloro-3-indolyl b-D-glucuronide, Cyclohexylammonium Salt, TRC

Product Code. 30330756
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
This item is not returnable. View return policy

Product Code. 30330756

Brand: Toronto Research Chemicals TRCB682380500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 5-Bromo-4-chloro-3-indolyl beta-D-glucuronide, Cyclohexylammonium salt
CAS 114162-64-0
Molecular Formula C14 H13 Br Cl N O7 . C6 H13 N
Quantity 500 mg
Synonym β-D-Glucopyranosiduronic acid, 5-bromo-4-chloro-1H-indol-3-yl, compd. with cyclohexanamine (1:1), X-Gluc, 5-Bromo-4-chloro-3-indolyl β-D-glucuronide Cyclohexylammonium Salt
InChI Formula InChI=1S/C14H13BrClNO7.C6H13N/c15-4-1-2-5-7(8(4)16)6(3-17-5)23-14-11(20)9(18)10(19)12(24-14)13(21)22;7-6-4-2-1-3-5-6/h1-3,9-12,14,17-20H,(H,21,22);6H,1-5,7H2/t9-,10-,11+,12-,14+;/m0./s1
SMILES NC1CCCCC1.O[C@@H]2[C@@H](O)[C@H](Oc3c[nH]c4ccc(Br)c(Cl)c34)O[C@@H]([C@H]2O)C(=O)O
IUPAC Name (2S,3S,4S,5R,6S)-6-[(5-bromo-4-chloro-1H-indol-3-yl)oxy]-3,4,5-trihydroxyoxane-2-carboxylic acid;cyclohexanamine
Molecular Weight (g/mol) 521.79
Formula Weight 520.0612
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.