missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-3-pyridineacetic acid, 98+%

Product Code. 11393147
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11393147 1 g 1g
11303157 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11393147 Supplier Thermo Scientific Alfa Aesar Supplier No. L13579.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 39891-12-8 | C7H6BrNO2 | 216.034 g/mol

5-Bromo-3-pyridineacetic acid acts as an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Bromo-3-pyridineacetic acid acts as an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff field.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39891-12-8
Molecular Formula C7H6BrNO2
Molecular Weight (g/mol) 216.034
MDL Number MFCD00829308
InChI Key UETIDNDXXGCJCE-UHFFFAOYSA-N
Synonym 5-bromo-3-pyridylacetic acid, 2-5-bromopyridin-3-yl acetic acid, 5-bromo-3-pyridineacetic acid, 3-pyridineacetic acid, 5-bromo, 2-5-bromo-3-pyridyl acetic acid, 5-bromo-3-pyridylaceticacid, 5-bromo-pyridin-3-yl-acetic acid, 5-bromopyridin-3-yl acetic acid, 5-bromo-3-pyridinyl acetic acid, 2-5-bromopyridin-3-yl acetic acid
PubChem CID 2802539
IUPAC Name 2-(5-bromopyridin-3-yl)acetic acid
SMILES C1=C(C=NC=C1Br)CC(=O)O

Specifications

Melting Point 182°C to 185°C
Quantity 1 g
Beilstein 388264
Solubility Information Soluble in water.
Formula Weight 216.03
Percent Purity ≥98%
Chemical Name or Material 5-Bromo-3-pyridineacetic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.