missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-3-nitropyridin-2-ol, 95%+, Thermo Scientific™

Product Code. 10124054
Change view
Click to view available options
Quantity:
10 g
25 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10124054 25 g Amber Glass Bottle 25g
10203482 10 g Amber Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10124054 Supplier Thermo Scientific Maybridge Supplier No. SPB06123EB
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 15862-34-7
Molecular Formula C5H3BrN2O3
Molecular Weight (g/mol) 218.994
InChI Key WXRLCVUDLFFTFF-UHFFFAOYSA-N
Synonym 5-bromo-2-hydroxy-3-nitropyridine, 5-bromo-3-nitropyridin-2-ol, 5-bromo-3-nitro-2-pyridinol, 5-bromo-3-nitro-2-pyridone, 5-bromo-3-nitropyridin-2 1h-one, 5-bromo-3-nitro-pyridin-2-ol, 3-bromo-6-hydroxy-5-nitropyridine, 5-bromo-3-nitro-2 1h-pyridinone, 5-bromo-3-nitro-2-hydroxypyridine, 2-hydroxy-3-nitro-5-bromopyridine
PubChem CID 85147
IUPAC Name 5-bromo-3-nitro-1H-pyridin-2-one
SMILES C1=C(C(=O)NC=C1Br)[N+](=O)[O-]

Specifications

Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 25 g
Formula Weight 218.99
Percent Purity 95%
Chemical Name or Material 5-Bromo-3-nitropyridin-2-ol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.