missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-3-methylpyridin-2-amine, 97%, Thermo Scientific™

Product Code. 10463024
Change view
Click to view available options
Quantity:
10 g
25 g
Unit Size:
10g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10463024 25 g 25g
10183624 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10463024 Supplier Thermo Scientific Maybridge Supplier No. XBX00112EB
 Restricted Item
Estimated Shipment: 29-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 3430-21-5
Molecular Formula C6H7BrN2
Molecular Weight (g/mol) 187.04
InChI Key KBLGGRWUEVCNPY-UHFFFAOYSA-N
Synonym 2-amino-5-bromo-3-methylpyridine, 2-amino-5-bromo-3-picoline, 2-amino-3-methyl-5-bromopyridine, 5-bromo-3-methyl-pyridin-2-ylamine, 2-pyridinamine, 5-bromo-3-methyl, 5-bromo-3-methyl-2-pyridinamine, 2-amina-5-bromo-3-picoline, 5-bromo-3-methyl-2-pyridylamine, 5-bromo-3-methyl-pyridin-2-amine, 5-bromo-3-methylpyridin-2-ylamine
PubChem CID 137934
IUPAC Name 5-bromo-3-methylpyridin-2-amine
SMILES CC1=CC(=CN=C1N)Br

Specifications

Assay Percent Range 97%
Quantity 25 g
Formula Weight 187.04
Percent Purity 97%
Chemical Name or Material 5-Bromo-3-methylpyridin-2-amine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.