missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-3-methyl-benzene-1,2-diamine, 97%, Thermo Scientific™

Product Code. 10524653
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10524653 1 g Amber Glass Bottle 1g
10422484 5 g Amber Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10524653 Supplier Thermo Scientific Maybridge Supplier No. MO07383DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 76153-06-5
Molecular Formula C7H9BrN2
Molecular Weight (g/mol) 201.067
InChI Key UOFSLKHZOPVGHG-UHFFFAOYSA-N
Synonym 5-bromo-3-methyl-benzene-1,2-diamine, 5-bromo-3-methyl-1,2-benzenediamine, 1,2-benzenediamine, 5-bromo-3-methyl, 2,3-diamino-5-bromotoluene, 5-bromo-2,3-diaminotoluene, bromomethylbenzenediamine, pubchem22667, ksc642k2n, 5-bromo-2,3-diamino-toluene, 5-bromo-2.3-diamino-toluene
PubChem CID 1516397
IUPAC Name 5-bromo-3-methylbenzene-1,2-diamine
SMILES CC1=CC(=CC(=C1N)N)Br

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 201.07
Percent Purity 97%
Chemical Name or Material 5-Bromo-3-methylbenzene-1, 2-diamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.