missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-3-methoxyquinoline, 96%

Product Code. 15491058
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15491058 1 g 1g
15411068 250 mg 250mg
15401068 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15491058 Supplier Thermo Scientific Alfa Aesar Supplier No. H54919.03
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 776296-12-9 | C10H8BrNO | 238.084 g/mol

It is an active pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an active pharmaceutical intermediate.

Notes
Refrigerated (2-4°C)
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 776296-12-9
Molecular Formula C10H8BrNO
Molecular Weight (g/mol) 238.084
MDL Number MFCD16038652
InChI Key YNVIAXULFXXTLV-UHFFFAOYSA-N
Synonym 5-bromo-3-methoxy-quinoline, 5-bromo-3-methyoxy-quinoline, quinoline, 5-bromo-3-methoxy
PubChem CID 58227851
IUPAC Name 5-bromo-3-methoxyquinoline
SMILES COC1=CN=C2C=CC=C(C2=C1)Br

Specifications

Quantity 1 g
Formula Weight 238.08
Percent Purity 96%
Chemical Name or Material 5-Bromo-3-methoxyquinoline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.