missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ 5-Bromo-3-fluoropyridine-2-carboxylic acid, 98%
Click to view available options
5g
Specifications
CAS | 669066-91-5 |
Melting Point | 175°C to 180°C |
Molecular Formula | C6H3BrFNO2 |
MDL Number | MFCD07437865 |
Quantity | 5 g |
Beilstein | 11103426 |
Synonym | 5-bromo-3-fluoropicolinic acid, 5-bromo-3-fluoro-2-pyridinecarboxylic acid, 2-pyridinecarboxylic acid, 5-bromo-3-fluoro, 5-bromo-3-fluoropicolinicacid, 5-bromo-2-carboxy-3-fluoropyridine, 5-bromo-3-fluoropyridin-2-carboxylicacid, 2-pyridinecarboxylicacid,5-bromo-3-fluoro, 2-pyridinecarboxylicacid, 5-bromo-3-fluoro, 5-bromo-3-fluoro-pyridine-2-carboxylic acid, 5-bromanyl-3-fluoranyl-pyridine-2-carboxylic acid |
InChI Key | JXHUXOAJAYWMHK-UHFFFAOYSA-N |
SMILES | OC(=O)C1=NC=C(Br)C=C1F |
IUPAC Name | 5-bromo-3-fluoropyridine-2-carboxylic acid |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'