missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-(trifluoromethyl)pyridine-4-carboxylic acid, 95%, Thermo Scientific Chemicals

Product Code. 15498045
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15498045 1 g 1g
15408055 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15498045 Supplier Thermo Scientific Alfa Aesar Supplier No. H32316.03
 Restricted Item
Estimated Shipment: 14-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 749875-16-9 | Molecular Formula: C7H3BrF3NO2 | Molecular Weight: 270.01 g/mol

5-Bromo-2-(trifluoromethyl)pyridine-4-carboxylic acid, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Bromo-2-(trifluoromethyl)pyridine-4-carboxylic acid, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Slightly soluble in water, soluble in organic solvents.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 749875-16-9
Molecular Formula C7H3BrF3NO2
Molecular Weight (g/mol) 270.01
MDL Number MFCD10687236
InChI Key XNVAVQMVAYUTBA-UHFFFAOYSA-N
Synonym 5-bromo-2-trifluoromethyl isonicotinic acid, 5-bromo-2-trifluoromethyl pyridine-4-carboxylic acid, 5-bromo-2-trifluoromethyl-isonicotinic acid, 4-pyridinecarboxylic acid, 5-bromo-2-trifluoromethyl, 5-bromo-2-trifluoromethyl isonicotinicacid, 3-bromo-6-trifluoromethyl pyridine-4-carboxylic acid
PubChem CID 28306936
IUPAC Name 5-bromo-2-(trifluoromethyl)pyridine-4-carboxylic acid
SMILES OC(=O)C1=CC(=NC=C1Br)C(F)(F)F

Specifications

Melting Point 235°C to 238°C
Quantity 1 g
Solubility Information Slightly soluble in water,soluble in organic solvents.
Formula Weight 270
Percent Purity 95%
Chemical Name or Material 5-Bromo-2-(trifluoromethyl)pyridine-4-carboxylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.