missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-thienylzinc bromide, 0.5M in THF, packaged under Argon in resealable ChemSeal™ bottles

Product Code. 15415435
Change view
Click to view available options
Quantity:
50 mL
Unit Size:
50mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15415435 50 mL 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15415435 Supplier Thermo Scientific Alfa Aesar Supplier No. H58942.AD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 151073-94-8 | C4H2Br2SZn | 307.308 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 151073-94-8
Molecular Formula C4H2Br2SZn
MDL Number MFCD00671986
Quantity 50 mL
UN Number UN3399
Sensitivity Air sensitive
Synonym 5-bromo-2-thienylzinc bromide, 5-bromo-2-thienylzinc bromide solution, bromo 5-bromothiophen-2-yl zinc, 5-bromo-2-thienylzinc bromide 0.5 m in tetrahydrofuran, 5-bromo-2-thienylzinc bromide, 0.5m in thf, packaged under argon in resealable chemseal bottles
InChI Key OSGCPOFIURDPFT-UHFFFAOYSA-M
SMILES C1=[C-]SC(=C1)Br.[Zn+]Br
IUPAC Name 5-bromo-2H-thiophen-2-ide;bromozinc(1+)
Molecular Weight (g/mol) 307.308
PubChem CID 4413080
Formula Weight 307.31
Chemical Name or Material 5-Bromo-2-thienylzinc bromide, Packaged under Argon in resealable ChemSeal™ bottles
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.