missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-pyridineacetic acid, 98%, Thermo Scientific Chemicals

Product Code. p-7031915
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15445977 250 mg 250mg
15435977 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15445977 Supplier Thermo Scientific Alfa Aesar Supplier No. H33181.MD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 192642-85-6
Molecular Formula C7H6BrNO2
Molecular Weight (g/mol) 216.03
MDL Number MFCD09999983
InChI Key ATKULCGQSLCGEK-UHFFFAOYSA-N
Synonym 2-5-bromopyridin-2-yl acetic acid, 5-bromopyridine-2-acetic acid, 5-bromopyridin-2-yl acetic acid, 2-pyridineacetic acid, 5-bromo, 5-bromo-2-pyridineacetic acid, 5-bromo-pyridin-2-yl-acetic acid, 5-bromo-pyridylacetic acid, acmc-209ew7, 5-bromo-2-pyridyl acetic acid, 5-bromo-2-pyridinyl acetic acid
PubChem CID 46238459
IUPAC Name 2-(5-bromopyridin-2-yl)acetic acid
SMILES OC(=O)CC1=NC=C(Br)C=C1

Specifications

Melting Point 120°C to 123°C
Quantity 250 mg
Formula Weight 216.03
Percent Purity 98%
Chemical Name or Material 5-Bromo-2-pyridineacetic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.