missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-pyrazinamine, Tech., Thermo Scientific™

Product Code. 10134204
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10134204 5 g 5g
10631573 1 g 1g
10392155 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10134204 Supplier Thermo Scientific Maybridge Supplier No. MO00943DE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 59489-71-3
Molecular Formula C4H4BrN3
Molecular Weight (g/mol) 174.001
MDL Number MFCD00235015
InChI Key KRRTXVSBTPCDOS-UHFFFAOYSA-N
Synonym 2-amino-5-bromopyrazine, 5-bromo-2-pyrazinamine, 5-bromo-pyrazin-2-ylamine, 2-bromo-5-aminopyrazine, pyrazinamine, 5-bromo, 5-amino-2-bromopyrazine, 5-bromo-2-aminopyrazine, 5-bromopyrazin-2-ylamine, 5-bromopyrazinamine, 2-amino-5-bromo-pyrazine
PubChem CID 599539
IUPAC Name 5-bromopyrazin-2-amine
SMILES C1=C(N=CC(=N1)Br)N

Specifications

CAS Min % 97.0
Packaging Amber glass bottle
Quantity 5 g
Formula Weight 174
Percent Purity Tech.
Grade Technical
Chemical Name or Material 5-Bromo-2-pyrazinamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.