missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-nitrobenzotrifluoride, 98%, Thermo Scientific™

Product Code. 10606681 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
Unit Size:
5g
This item is not returnable. View return policy

Product Code. 10606681

Brand: Thermo Scientific Acros 308890050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 344-38-7
CAS Min % 97.5
CAS Max % 100.0
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C7H3BrF3NO2
Linear Formula BrC6H3(NO)CF3
Refractive Index 1.5220 to 1.5240 (20°C, 589nm)
MDL Number MFCD00014706
Quantity 5 g
Synonym 5-bromo-2-nitrobenzotrifluoride, 4-bromo-1-nitro-2-trifluoromethyl benzene, 2-nitro-5-bromobenzotrifluoride, 5-bromo-2-nitro-a,a,a-trifluorotoluene, 5-bromo-a-a,a-trifluoro-2-nitrotoluene, 4-bromo-1-nitro-2 trifluoromethyl benzene, 4-bromo-1-nitro-2-trifluoromethyl-benzene, benzene, 4-bromo-1-nitro-2-trifluoromethyl, 5-bromo-alpha,alpha,alpha-trifluoro-2-nitrotoluene, 2-nitro-5-bromotrifluorotoluol
InChI Key ZHLYHEDQTJZYFI-UHFFFAOYSA-N
SMILES C1=CC(=C(C=C1Br)C(F)(F)F)[N+](=O)[O-]
IUPAC Name 4-bromo-1-nitro-2-(trifluoromethyl)benzene
Molecular Weight (g/mol) 270.005
PubChem CID 136171
Formula Weight 270.01
Percent Purity ≥97.5%
Physical Form Liquid
Chemical Name or Material 5-Bromo-2-nitrobenzotrifluoride, 98%
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.