Learn More
5-Bromo-2-(methylsulfonyl)pyrimidine, 97%, ACROS Organics™
Brand: Acros Organics 432390050
Code : 29
Additional Details : CAS Number : 38275-48-8 Weight : 0.05500kg
Quantity | 5g |
---|---|
Packaging | Glass bottle |
Chemical Identifiers
5-Bromo-2-(methylsulfonyl)pyrimidine | |
96.0 | |
≥96.0% | |
C5H5BrN2O2S | |
MFCD00508247 |
38275-48-8 | |
100.0 | |
97% | |
237.08 |
Specifications
5-Bromo-2-(methylsulfonyl)pyrimidine | |
96.0 | |
Authentic | |
130° to 134°C | |
Crystalline Powder and/or Needles | |
≥96.0% | |
C5H5BrN2O2S | |
MFCD00508247 |
38275-48-8 | |
100.0 | |
Glass bottle | |
White | |
5g | |
97% | |
237.08 |