missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-methylpyrimidine, 97%, Thermo Scientific™

Product Code. 15294784 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Unit Size:
1g
This item is not returnable. View return policy

Product Code. 15294784

Brand: Thermo Scientific Acros 449880010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 7752-78-5
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Molecular Formula C5H5BrN2
Quantity 1 g
Synonym 5-bromo-2-methyl-pyrimidine, 2-methyl-5-bromopyrimidine, pyrimidine, 5-bromo-2-methyl, 5-bromo-2-methyl-1,3-diazine, pubchem15231, acmc-1bm3h, 5-bromo-2-methyl pyrimidine
InChI Key NEDJTEXNSTUKHW-UHFFFAOYSA-N
SMILES CC1=NC=C(C=N1)Br
IUPAC Name 5-bromo-2-methylpyrimidine
Molecular Weight (g/mol) 173.01
PubChem CID 14387744
Formula Weight 173.01
Percent Purity 97%
Chemical Name or Material 5-Bromo-2-methylpyrimidine
Show More Show Less
compliance-icons
Product Identifier
  • 5-Bromo-2-methylpyrimidine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.