missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-methylphenol, 95%

Product Code. p-7029489
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
This item is not returnable. View return policy

Product Code. 11451560

Brand: Thermo Scientific Alfa Aesar H26956.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 36138-76-8 | C7H7BrO | 187.036 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 36138-76-8
Molecular Formula C7H7BrO
Molecular Weight (g/mol) 187.036
MDL Number MFCD08061906
InChI Key OONJCAWRVJDVBB-UHFFFAOYSA-N
Synonym 5-bromo-o-cresol, phenol, 5-bromo-2-methyl, 4-bromo-2-hydroxytoluene, 5-bromo-2-methyl phenol, 5-bromo-2-methyl-phenol, 3-bromo-6-methylphenol, 2-hydroxy-4-bromotoluene, 5-brom-2-kresol, zlchem 468, pubchem4132
PubChem CID 12648404
IUPAC Name 5-bromo-2-methylphenol
SMILES CC1=C(C=C(C=C1)Br)O

Specifications

Melting Point 77°C to 80°C
Quantity 1 g
Formula Weight 187.04
Percent Purity 95%
Chemical Name or Material 5-Bromo-2-methylphenol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.