missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-methylbenzoic acid, 97%, Thermo Scientific Chemicals

Catalog Number 15466988
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
79669-49-1
C8H7BrO2
215.05
MFCD00267350
SEENCYZQHCUTSB-UHFFFAOYSA-N
2-methyl-5-bromobenzoic acid, 5-bromo-o-toluic acid, 5-bromo-2-methyl-benzoic acid, 5-bromo-2-methyl benzoic acid, synquest 2721-9-x8, benzoic acid, 5-bromo-2-methyl, pubchem4728, acmc-1bhwo, 3-bromo-6-methylbenzoic acid, ksc497q7f
346004
5-bromo-2-methylbenzoic acid
CC1=CC=C(Br)C=C1C(O)=O
This item is not returnable. View return policy
79669-49-1
C8H7BrO2
215.05
MFCD00267350
SEENCYZQHCUTSB-UHFFFAOYSA-N
2-methyl-5-bromobenzoic acid, 5-bromo-o-toluic acid, 5-bromo-2-methyl-benzoic acid, 5-bromo-2-methyl benzoic acid, synquest 2721-9-x8, benzoic acid, 5-bromo-2-methyl, pubchem4728, acmc-1bhwo, 3-bromo-6-methylbenzoic acid, ksc497q7f
346004
5-bromo-2-methylbenzoic acid
CC1=CC=C(Br)C=C1C(O)=O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 167°C to 171°C
Quantity 1 g
Formula Weight 215.05
Percent Purity 97%
Chemical Name or Material 5-Bromo-2-methylbenzoic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'