missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-methylaniline, 98%, Thermo Scientific Chemicals

Product Code. 10135312 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10135312 25 g Glass bottle 25g
10598533 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10135312 Supplier Thermo Scientific Acros Supplier No. 372710250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39478-78-9
MDL Number MFCD00800678
InChI Key RXQNKKRGJJRMKD-UHFFFAOYSA-N
Synonym 2-amino-4-bromotoluene, 5-bromo-o-toluidine, 3-bromo-6-methylaniline, 5-bromo-2-methyl-aniline, benzenamine, 5-bromo-2-methyl, 5-bromo-2-methyl aniline, 5-bromo-2-methyl-phenylamine, zlchem 583, pubchem3801, acmc-1ajfs
PubChem CID 2734805
IUPAC Name 5-bromo-2-methylaniline
SMILES CC1=C(C=C(C=C1)Br)N

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Yellow to Brown
Density 1.4900g/mL
Boiling Point 139°C
Flash Point 110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Molecular Formula C7H8BrN
Linear Formula BrC6H3(CH3)NH2
Refractive Index 1.6120 to 1.614
Quantity 25 g
Specific Gravity 1.49
Molecular Weight (g/mol) 186.05
Formula Weight 186.05
Percent Purity 98%
Physical Form Liquid After Melting
Chemical Name or Material 5-Bromo-2-methylaniline
Show More Show Less
compliance-icons
Product Identifier
  • 5-Bromo-2-methylaniline
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.