missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-methyl-4-nitro-1H-imidazole, 97%, Thermo Scientific™

Product Code. 10496832
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10496832 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10496832 Supplier Thermo Scientific Maybridge Supplier No. BTB09770DA
 Restricted Item
Estimated Shipment: 10-02-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 18874-52-7
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C4H4BrN3O2
MDL Number MFCD00156130
Quantity 1 g
Synonym 4-bromo-2-methyl-5-nitro-1h-imidazole, 5-bromo-2-methyl-4-nitroimidazole, 4-bromo-2-methyl-5-nitro-3h-imidazole, 4-bromo-2-methyl-5-nitroimidazole, 1h-imidazole, 4-bromo-2-methyl-5-nitro, 2-methyl-4-nitro-5-bromoimidazole, imidazole, 5-bromo-4-nitro-2-methyl, 2-methyl-4-bromo-5-nitro-1h-imidazole, 1h-imidazole,5-bromo-2-methyl-4-nitro, 1h-imidazole, 5-bromo-2-methyl-4-nitro
InChI Key YOJYWZSEUWUYAQ-UHFFFAOYSA-N
SMILES CC1=NC(=C(N1)Br)[N+](=O)[O-]
IUPAC Name 5-bromo-2-methyl-4-nitro-1H-imidazole
Molecular Weight (g/mol) 205.999
PubChem CID 29322
Formula Weight 206
Percent Purity 97%
Chemical Name or Material 5-Bromo-2-methyl-4-nitro-1H-imidazole
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.