missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-methyl-3-nitropyridine, 96%, Thermo Scientific Chemicals

Product Code. p-7036599
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 15481058

Brand: Thermo Scientific Alfa Aesar H54917.MD

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 911434-05-4
Molecular Formula C6H5BrN2O2
Molecular Weight (g/mol) 217.022
MDL Number MFCD09031419
InChI Key FZZLWWNOYMHSIS-UHFFFAOYSA-N
Synonym 5-bromo-3-nitropicoline, 5-bromo-3-nitro-2-picoline, 5-bromo-2-methyl-3-nitro-pyridine, pyridine, 5-bromo-2-methyl-3-nitro, pubchem13809, ksc496e9t, bromo-2-methyl-3-nitropyridine
PubChem CID 25067266
IUPAC Name 5-bromo-2-methyl-3-nitropyridine
SMILES CC1=NC=C(C=C1[N+](=O)[O-])Br

Specifications

Quantity 250 mg
Formula Weight 217.02
Percent Purity 96%
Chemical Name or Material 5-Bromo-2-methyl-3-nitropyridine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.