missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-methyl-2-pentene, 97%

Product Code. 11318706
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11318706 1 g 1g
11328706 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11318706 Supplier Thermo Scientific Alfa Aesar Supplier No. L19406.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2270-59-9 | C6H11Br | 163.06 g/mol

5-Bromo-2-methyl-2-pentene acts as a synthetic reagent, which is useful in the formation of sesqueterpenoid. It is also employed in the preparation of geranlol-3-14C. Further, it is used in the preparation of penifulvin A, which is a sesquiterpenoid with a novel dioxa-fenestrane structure.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Bromo-2-methyl-2-pentene acts as a synthetic reagent, which is useful in the formation of sesqueterpenoid. It is also employed in the preparation of geranlol-3-14C. Further, it is used in the preparation of penifulvin A, which is a sesquiterpenoid with a novel dioxa-fenestrane structure.

Solubility
Slightly miscible with water.

Notes
Store in a cool place. Incompatible with strong oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2270-59-9
Molecular Formula C6H11Br
Molecular Weight (g/mol) 163.06
MDL Number MFCD00009887
InChI Key UNXURIHDFUQNOC-UHFFFAOYSA-N
Synonym 5-bromo-2-methyl-2-pentene, acmc-20ah0m, 1-bromo4-methyl-3-pentene, 5-brom-2-methyl-2-pentene, 1-bromo-4-methyl-3-pentene, 2-methyl-5-bromo-2-pentene, 2-pentene,5-bromo-2-methyl, 5-bromo-2-methyl-pent-2-ene
PubChem CID 137521
IUPAC Name 5-bromo-2-methylpent-2-ene
SMILES CC(C)=CCCBr

Specifications

Density 1.219
Boiling Point 50°C (20 mmHg)
Flash Point 23°C (73°F)
Refractive Index 1.4775
Quantity 1 g
UN Number UN1993
Beilstein 1735749
Solubility Information Slightly miscible with water.
Formula Weight 163.06
Percent Purity 97%
Chemical Name or Material 5-Bromo-2-methyl-2-pentene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.