missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-methoxypyridine, 97%

Product Code. 10280153 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
Packaging:
Glass bottle
Unit Size:
25mL
5mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10280153 5 mL Glass bottle 5mL
10143982 25 mL Glass bottle 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10280153 Supplier Thermo Scientific Acros Supplier No. 376880050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 13472-85-0 | C6H6BrNO | 188.024 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13472-85-0
MDL Number MFCD01318952
InChI Key XADICJHFELMBGX-UHFFFAOYSA-N
Synonym 2-methoxy-5-bromopyridine, pyridine, 5-bromo-2-methoxy, 3-bromo-6-methoxypyridine, 5-bromo-2-methoxy-pyridine, 2-methyloxy-5-bromopyridine, 5-bromo-2-methoxy pyridine, 2-methoxy-5-bromo-pyridine, 2-methyloxy-5-bromo pyridine, zlchem 355, pubchem2408
PubChem CID 2734895
IUPAC Name 5-bromo-2-methoxypyridine
SMILES COC1=NC=C(C=C1)Br

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.4530g/mL
Boiling Point 80.0°C (12.0 mmHg)
Flash Point 96°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Molecular Formula C6H6BrNO
Refractive Index 1.5540 to 1.5560
Quantity 5 mL
Specific Gravity 1.453
Molecular Weight (g/mol) 188.024
Formula Weight 188.02
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 5-Bromo-2-methoxypyridine
Show More Show Less
compliance-icons
Product Identifier
  • 5-Bromo-2-methoxypyridine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.