missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-methoxypyridine-3-carboxaldehyde, 99%, Thermo Scientific™

Product Code. 11464460
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11464460 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11464460 Supplier Thermo Scientific Alfa Aesar Supplier No. H50062.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
5-Bromo-2-methoxypyridine-3-carboxaldehyde is used as a synthetic building block. It is also used as an pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Light and moisture sensitive. Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103058-87-3
Molecular Formula C7H6BrNO2
Molecular Weight (g/mol) 216.03
MDL Number MFCD05663521
InChI Key VRNOWEKCASDTFG-UHFFFAOYSA-N
Synonym 5-bromo-2-methoxy-pyridine-3-carbaldehyde, 5-bromo-2-methoxynicotinaldehyde, 5-bromo-2-methoxypyridine-3-carboxaldehyde, 3-pyridinecarboxaldehyde, 5-bromo-2-methoxy, 5-bromo-2-methoxypyridin-3-carboxaldehyde, 5-bromo-2-methoxy-3-pyridinecarboxaldehyde, pubchem9207, abbypharma ap-10-5904, 3-pyridinecarboxaldehyde,5-bromo-2-methoxy, 5-bromo-2-methoxypyridine-3-carboxaldehyde 1g
PubChem CID 1516403
IUPAC Name 5-bromo-2-methoxypyridine-3-carbaldehyde
SMILES COC1=NC=C(Br)C=C1C=O

Specifications

Quantity 5 g
Sensitivity Air Sensitive
Solubility Information Slightly soluble in water.
Formula Weight 216.04
Percent Purity 99%
Chemical Name or Material 5-Bromo-2-methoxypyridine-3-carboxaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.