missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-methoxybenzaldehyde, 98+%

Product Code. 11423997
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11423997 25 g 25g
11433997 100 g 100g
11443997 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11423997 Supplier Thermo Scientific Alfa Aesar Supplier No. A19396.14
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 25016-01-7 | C8H7BrO2 | 215.046 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25016-01-7
Molecular Formula C8H7BrO2
Molecular Weight (g/mol) 215.046
MDL Number MFCD00003347
InChI Key IJIBRSFAXRFPPN-UHFFFAOYSA-N
Synonym 5-bromo-2-anisaldehyde, 5-bromo-o-anisaldehyde, 2-methoxy-5-bromobenzaldehyde, benzaldehyde, 5-bromo-2-methoxy, 5-bromo-2-methoxy-benzaldehyde, pubchem2644, acmc-209ggg, intermediates-zcf02672, o-anisaldehyde, 5-bromo, ksc494q2j
PubChem CID 90684
IUPAC Name 5-bromo-2-methoxybenzaldehyde
SMILES COC1=C(C=C(C=C1)Br)C=O

Specifications

Melting Point 114°C to 118°C
Quantity 25 g
Beilstein 508840
Sensitivity Air Sensitive
Formula Weight 215.05
Percent Purity ≥98%
Chemical Name or Material 5-Bromo-2-methoxybenzaldehyde

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.