missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-isopropoxybenzonitrile, 97%, Thermo Scientific Chemicals

Product Code. 11400150
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11400150 1 g 1g
11490140 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11400150 Supplier Thermo Scientific Alfa Aesar Supplier No. H50627.03
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
This item is not returnable. View return policy

5-Bromo-2-isopropoxybenzonitrile is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Bromo-2-isopropoxybenzonitrile is used as pharmaceutical intermediate.

Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 515832-52-7
Molecular Formula C10H10BrNO
Molecular Weight (g/mol) 240.1
MDL Number MFCD02257441
InChI Key YRPPYKWHWKFYGJ-UHFFFAOYSA-N
Synonym 5-bromo-2-isopropoxybenzonitrile, 5-bromo-2-isopropoxy-benzonitrile, benzonitrile, 5-bromo-2-1-methylethoxy, 5-bromo-2-propan-2-yloxy benzonitrile, 5-bromo-2-methylethoxy benzenecarbonitrile, acmc-1awgy, 5-bromo-2 1-methylethyl oxy benzonitrile, 5-bromo-2-1-methylethyl oxy benzonitrile
PubChem CID 3262494
IUPAC Name 5-bromo-2-propan-2-yloxybenzonitrile
SMILES CC(C)OC1=C(C=C(C=C1)Br)C#N

Specifications

Quantity 1 g
UN Number UN3439
Formula Weight 240.1
Percent Purity 97%
Chemical Name or Material 5-Bromo-2-isopropoxybenzonitrile

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.