missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-hydroxybenzyl alcohol, 98+%, Thermo Scientific™

Product Code. 11419473
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11419473

Brand: Thermo Scientific Alfa Aesar A12344.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2316-64-5
Molecular Formula C7H7BrO2
Molecular Weight (g/mol) 203.04
MDL Number MFCD00004618
InChI Key KNKRHSVKIORZQB-UHFFFAOYSA-N
Synonym 5-bromo-2-hydroxybenzyl alcohol, 4-bromo-2-hydroxymethyl phenol, bromosaligenin, 5-bromosalicyl alcohol, bromsalizol, 5-bromosaligenin, 5-bromo-2-hydroxybenzylalcohol, 2-hydroxymethyl-4-bromophenol, benzenemethanol, 5-bromo-2-hydroxy, unii-y1uz53815e
PubChem CID 75342
IUPAC Name 4-bromo-2-(hydroxymethyl)phenol
SMILES OCC1=CC(Br)=CC=C1O

Specifications

Melting Point 108°C to 110°C
Quantity 100 g
Beilstein 509726
Formula Weight 203.04
Percent Purity ≥98%
Chemical Name or Material 5-Bromo-2-hydroxybenzyl alcohol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.