missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-hydroxybenzyl alcohol, 98%, Thermo Scientific Chemicals

Product Code. 10317801
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2316-64-5
Molecular Formula C7H7BrO2
Molecular Weight (g/mol) 203.04
MDL Number MFCD00004618
InChI Key KNKRHSVKIORZQB-UHFFFAOYSA-N
Synonym 5-bromo-2-hydroxybenzyl alcohol, 4-bromo-2-hydroxymethyl phenol, bromosaligenin, 5-bromosalicyl alcohol, bromsalizol, 5-bromosaligenin, 5-bromo-2-hydroxybenzylalcohol, 2-hydroxymethyl-4-bromophenol, benzenemethanol, 5-bromo-2-hydroxy, unii-y1uz53815e
PubChem CID 75342
IUPAC Name 4-bromo-2-(hydroxymethyl)phenol
SMILES OCC1=CC(Br)=CC=C1O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 106.0°C to 111.0°C
Color White to Brown
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula BrC6H3(OH)CH2OH
Quantity 5 g
Beilstein 06, 893
Merck Index 12, 1460
Solubility Information Solubility in water: insoluble in water. Other solubilities: soluble in acetone,benzene,chloroform
Formula Weight 203.04
Percent Purity 98%
Physical Form Crystalline Powder or Flakes
Chemical Name or Material 5-Bromo-2-hydroxybenzyl alcohol, 98%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'