missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-hydroxybenzeneboronic acid, 96%

Product Code. p-7035223
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 15439707

Brand: Thermo Scientific Alfa Aesar H52937.MD

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 89598-97-0 | C6H6BBrO3 | 216.825 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 89598-97-0
Molecular Formula C6H6BBrO3
Molecular Weight (g/mol) 216.825
MDL Number MFCD00093408
InChI Key XSXTWLOHAGPBNO-UHFFFAOYSA-N
Synonym 5-bromo-2-hydroxyphenyl boronic acid, 2-borono-4-bromophenol, 5-bromo-2-hydroxy benzeneboronic acid, benzeneboronic acid, 5-bromo-2-hydroxy, acmc-209r1x, 5-bromo-2-hydroxyphenyl boranediol, 5-bromo-2-hydroxybenzeneboronic acid, benzeneboronic acid,5-bromo-2-hydroxy-6ci,7ci
PubChem CID 44118764
IUPAC Name (5-bromo-2-hydroxyphenyl)boronic acid
SMILES B(C1=C(C=CC(=C1)Br)O)(O)O

Specifications

Melting Point 196°C to 200°C
Quantity 250 mg
Formula Weight 216.83
Percent Purity 97%
Chemical Name or Material 5-Bromo-2-hydroxybenzeneboronic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.