missing translation for 'onlineSavingsMsg'
Learn More

5'-Bromo-2'-hydroxyacetophenone, 98%

Product Code. 15438578
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15438578 50 g 50g
15428578 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15438578 Supplier Thermo Scientific Alfa Aesar Supplier No. H61979.18
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1450-75-5 | C8H7BrO2 | 215.046 g/mol

5?-Bromo-2?-hydroxyacetophenone (5-Bromo-2-hydroxyacetophenone) may be used in ligand in the preparation of tetrahedral metallocene complexes containing vanadium(IV), synthesis of {2?-[1-(5-bromo-2-oxidophenyl) ethylidene] benzohydrazidato (2-)} tris(pyridine) nickel(II)] pyridine solvate, synthesis of N?-[1-(5-bromo-2-hydroxyphenyl)ethylidene]-3,4,5-trihydroxybenzohydrazide dimethyl sulfoxide solvate trihydrate, preparation of 6-bromochromen-4-one.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5′-Bromo-2′-hydroxyacetophenone (5-Bromo-2-hydroxyacetophenone) may be used in ligand in the preparation of tetrahedral metallocene complexes containing vanadium(IV), synthesis of {2′-[1-(5-bromo-2-oxidophenyl) ethylidene] benzohydrazidato (2-)} tris(pyridine) nickel(II)] pyridine solvate, synthesis of N′-[1-(5-bromo-2-hydroxyphenyl)ethylidene]-3,4,5-trihydroxybenzohydrazide dimethyl sulfoxide solvate trihydrate, preparation of 6-bromochromen-4-one.

Solubility
Soluble in Chloroform

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1450-75-5
Molecular Formula C8H7BrO2
Molecular Weight (g/mol) 215.046
MDL Number MFCD00191850
InChI Key HQCCNFFIOWYINW-UHFFFAOYSA-N
Synonym 5'-bromo-2'-hydroxyacetophenone, 1-5-bromo-2-hydroxyphenyl ethanone, 2-acetyl-4-bromophenol, 5-bromo-2-hydroxyacetophenone, 2'-hydroxy-5'-bromoacetophenone, 1-5-bromo-2-hydroxyphenyl ethan-1-one, ethanone, 1-5-bromo-2-hydroxyphenyl, 2-hydroxy-5-bromoacetophenone, 1-5-bromo-2-hydroxyphenyl-ethanon
PubChem CID 95991
IUPAC Name 1-(5-bromo-2-hydroxyphenyl)ethanone
SMILES CC(=O)C1=C(C=CC(=C1)Br)O

Specifications

Melting Point 56°C to 60°C
Color White to Yellow
Quantity 50 g
Solubility Information Soluble in Chloroform
Formula Weight 215.04
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 5'-Bromo-2'-hydroxyacetophenone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.