missing translation for 'onlineSavingsMsg'
Learn More

5'-Bromo-2'-hydroxyacetophenone, 98%

Product Code. 15428578
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
This item is not returnable. View return policy

Product Code. 15428578

Brand: Thermo Scientific Alfa Aesar H61979.09

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1450-75-5 | C8H7BrO2 | 215.046 g/mol

5?-Bromo-2?-hydroxyacetophenone (5-Bromo-2-hydroxyacetophenone) may be used in ligand in the preparation of tetrahedral metallocene complexes containing vanadium(IV), synthesis of {2?-[1-(5-bromo-2-oxidophenyl) ethylidene] benzohydrazidato (2-)} tris(pyridine) nickel(II)] pyridine solvate, synthesis of N?-[1-(5-bromo-2-hydroxyphenyl)ethylidene]-3,4,5-trihydroxybenzohydrazide dimethyl sulfoxide solvate trihydrate, preparation of 6-bromochromen-4-one.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5′-Bromo-2′-hydroxyacetophenone (5-Bromo-2-hydroxyacetophenone) may be used in ligand in the preparation of tetrahedral metallocene complexes containing vanadium(IV), synthesis of {2′-[1-(5-bromo-2-oxidophenyl) ethylidene] benzohydrazidato (2-)} tris(pyridine) nickel(II)] pyridine solvate, synthesis of N′-[1-(5-bromo-2-hydroxyphenyl)ethylidene]-3,4,5-trihydroxybenzohydrazide dimethyl sulfoxide solvate trihydrate, preparation of 6-bromochromen-4-one.

Solubility
Soluble in Chloroform

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1450-75-5
Molecular Formula C8H7BrO2
Molecular Weight (g/mol) 215.046
MDL Number MFCD00191850
InChI Key HQCCNFFIOWYINW-UHFFFAOYSA-N
Synonym 5'-bromo-2'-hydroxyacetophenone, 1-5-bromo-2-hydroxyphenyl ethanone, 2-acetyl-4-bromophenol, 5-bromo-2-hydroxyacetophenone, 2'-hydroxy-5'-bromoacetophenone, 1-5-bromo-2-hydroxyphenyl ethan-1-one, ethanone, 1-5-bromo-2-hydroxyphenyl, 2-hydroxy-5-bromoacetophenone, 1-5-bromo-2-hydroxyphenyl-ethanon
PubChem CID 95991
IUPAC Name 1-(5-bromo-2-hydroxyphenyl)ethanone
SMILES CC(=O)C1=C(C=CC(=C1)Br)O

Specifications

Melting Point 56°C to 60°C
Color White to Yellow
Quantity 10 g
Solubility Information Soluble in Chloroform
Formula Weight 215.04
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 5'-Bromo-2'-hydroxyacetophenone

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.