missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-furoic acid, 99%

Product Code. 11482607
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11482607 50 g 50g
11492607 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11482607 Supplier Thermo Scientific Alfa Aesar Supplier No. A14840.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 585-70-6 | C5H3BrO3 | 190.98 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 585-70-6
Molecular Formula C5H3BrO3
Molecular Weight (g/mol) 190.98
MDL Number MFCD00003239
InChI Key YVTQHZDUDUCGRD-UHFFFAOYSA-N
Synonym 5-bromo-2-furoic acid, 5-bromofuroic acid, 5-bromo-furan-2-carboxylic acid, 5-bromo-2-furancarboxylic acid, 2-furancarboxylic acid, 5-bromo, 5-bromo-2-furanecarboxylic acid, 5-bromo furan-2-carboxylic acid, 2-furoic acid, 5-bromo, t5oj bvq ee, 5-bromofuroic?acid
PubChem CID 68511
IUPAC Name 5-bromofuran-2-carboxylic acid
SMILES C1=C(OC(=C1)Br)C(=O)O

Specifications

Melting Point 188°C to 192°C
Quantity 50 g
Beilstein 118354
Formula Weight 190.99
Percent Purity 99%
Chemical Name or Material 5-Bromo-2-furoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.