missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-furaldehyde, 98%, stab. with 2% ethanol

Product Code. 11465097
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11465097 50 g 50g
11455097 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11465097 Supplier Thermo Scientific Alfa Aesar Supplier No. B22536.18
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1899-24-7 | C5H3BrO2 | 174.98 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1899-24-7
Molecular Formula C5H3BrO2
Molecular Weight (g/mol) 174.98
MDL Number MFCD00159501
InChI Key WJTFHWXMITZNHS-UHFFFAOYSA-N
Synonym 5-bromo-2-furaldehyde, 5-bromofurfural, 5-bromo-2-furfural, 5-bromofuran-2-carboxaldehyde, 5-bromofurancarboxaldehyde, 2-furancarboxaldehyde, 5-bromo, 5-bromofuraldehyde, 5-bromo-furan-2-carbaldehyde, 2-furaldehyde, 5-bromo, 5-bromo-furfural
PubChem CID 600328
IUPAC Name 5-bromofuran-2-carbaldehyde
SMILES BrC1=CC=C(O1)C=O

Specifications

Melting Point 82°C to 85°C
Boiling Point 112°C (6 mmHg)
Quantity 50 g
Beilstein 108403
Sensitivity Air Sensitive
Formula Weight 174.98
Percent Purity 98%
Chemical Name or Material 5-Bromo-2-furaldehyde, stabilized with 2% ethanol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.