missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Bromo-2-fluorotoluene, 98+%, Thermo Scientific™
Beskrivelse
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
5-Bromo-2-fluorotoluene is used in the preparation of 5-bromo-2-fluorobenzyl bromide via N-bromosuccinimide (NBS) bromination, required for the synthesis of crown bromofluorobenzene and 3-(4-fuoro-3-methylphenyl)acrylamide. It is also used as an intermediates of medicine.
Solubility
Not miscible or difficult to mix in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
5-Bromo-2-fluorotoluene is used in the preparation of 5-bromo-2-fluorobenzyl bromide via N-bromosuccinimide (NBS) bromination, required for the synthesis of crown bromofluorobenzene and 3-(4-fuoro-3-methylphenyl)acrylamide. It is also used as an intermediates of medicine.
Solubility
Not miscible or difficult to mix in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Tekniske data
Tekniske data
| Density | 1.486 |
| Boiling Point | 94°C to 95°C (50 mmHg) |
| Flash Point | 73°C (163°F) |
| Refractive Index | 1.529 |
| Quantity | 5 g |
| Beilstein | 2242693 |
| Solubility Information | Not miscible or difficult to mix in water. |
| Formula Weight | 189.03 |
| Percent Purity | ≥98% |
| Chemical Name or Material | 5-Bromo-2-fluorotoluene |
RUO – Research Use Only
Produkttitel
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?