missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-fluorotoluene, 98+%, Thermo Scientific™

Product Code. 11459582
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11459582 100 g 100g
11439582 5 g 5g
11449582 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11459582 Supplier Thermo Scientific Alfa Aesar Supplier No. A10721.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
5-Bromo-2-fluorotoluene is used in the preparation of 5-bromo-2-fluorobenzyl bromide via N-bromosuccinimide (NBS) bromination, required for the synthesis of crown bromofluorobenzene and 3-(4-fuoro-3-methylphenyl)acrylamide. It is also used as an intermediates of medicine.

Solubility
Not miscible or difficult to mix in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51437-00-4
Molecular Formula C7H6BrF
Molecular Weight (g/mol) 189.027
MDL Number MFCD00000343
InChI Key VXKYOKPNAXNAFU-UHFFFAOYSA-N
Synonym 5-bromo-2-fluorotoluene, 2-fluoro-5-bromotoluene, benzene, 4-bromo-1-fluoro-2-methyl, 4-bromo-1-fluoro-2-methyl-benzene, pubchem1599, acmc-1asrc, 2-fluoro-5-bromo-toluene, 5-bromo-2-fluoro toluene, 5-bromo-2-fluoro-toluene, ksc274o3j
PubChem CID 123527
IUPAC Name 4-bromo-1-fluoro-2-methylbenzene
SMILES CC1=C(C=CC(=C1)Br)F

Specifications

Density 1.486
Boiling Point 94°C to 95°C (50 mmHg)
Flash Point 73°C (163°F)
Refractive Index 1.529
Quantity 100 g
Beilstein 2242693
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 189.03
Percent Purity ≥98%
Chemical Name or Material 5-Bromo-2-fluorotoluene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.