missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-fluorobenzotrifluoride, 98%, Thermo Scientific™

Product Code. 11442748
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11442748 25 g 25g
11432748 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11442748 Supplier Thermo Scientific Alfa Aesar Supplier No. A16736.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Pharmaceutical Intermediates.

Solubility
Soluble in water 700 g/L (20°C)

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 393-37-3
Molecular Formula C7H3BrF4
Molecular Weight (g/mol) 243.00
MDL Number MFCD00042493
InChI Key AJLIJYGWAXPEOK-UHFFFAOYSA-N
Synonym 5-bromo-2-fluorobenzotrifluoride, 4-bromo-1-fluoro-2-trifluoromethyl benzene, 2-fluoro-5-bromobenzotrifluoride, 4-fluoro-3-trifluoromethyl bromobenzene, 5-bromo-alpha,alpha,alpha,2-tetrafluorotoluene, 5-bromo-2-fluoro-trifluoromethyl benzene, 2-fluoro-5-bromo benzotrifluoride, 1-bromo-4-fluoro-3-trifluoromethyl benzene, benzene, 4-bromo-1-fluoro-2-trifluoromethyl, acmc-209j4s
PubChem CID 2736325
IUPAC Name 4-bromo-1-fluoro-2-(trifluoromethyl)benzene
SMILES FC1=CC=C(Br)C=C1C(F)(F)F

Specifications

Density 1.72
Boiling Point 79°C to 80°C (50 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.46
Quantity 25 g
Beilstein 2643550
Solubility Information Soluble in water 700g/L (20°C)
Formula Weight 243
Percent Purity 98%
Chemical Name or Material 5-Bromo-2-fluorobenzotrifluoride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.