missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-fluoro-m-xylene, 97%

Product Code. 11430030
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11430030 1 g 1g
11440030 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11430030 Supplier Thermo Scientific Alfa Aesar Supplier No. H50564.03
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 99725-44-7 | C8H8BrF | 203.054 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99725-44-7
Molecular Formula C8H8BrF
Molecular Weight (g/mol) 203.054
MDL Number MFCD01320701
InChI Key ZXPHUVHMBKRRJF-UHFFFAOYSA-N
Synonym 5-bromo-2-fluoro-m-xylene, 5-bromo-2-fluoro-3-xylene, 4-bromo-2,6-dimethylfluorobenzene, 1-bromo-4-fluoro-3,5-dimethylbenzene, 5-bromo-2-fluoro-1,3-xylene, benzene, 5-bromo-2-fluoro-1,3-dimethyl, 3,5-dimethyl-4-fluorobromobenzene, 5-bromo-2-fluoro-1,3-dimethyl-benzene, ksc486i9j
PubChem CID 2736298
IUPAC Name 5-bromo-2-fluoro-1,3-dimethylbenzene
SMILES CC1=CC(=CC(=C1F)C)Br

Specifications

Quantity 1 g
Boiling Point 95°C (22 mmHg)
Formula Weight 203.06
Percent Purity 97%
Chemical Name or Material 5-Bromo-2-fluoro-m-xylene
compliance-icons
Product Identifier
  • 5-Bromo-2-fluoro-1,3-dimethylbenzene
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: 5-Bromo-2-fluoro-1,3-dimethylbenzene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.