missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-fluoro-4-methylpyridine, 96%, Thermo Scientific™

Product Code. 15467978
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15467978 100 g 100g
15447978 5 g 5g
15457978 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15467978 Supplier Thermo Scientific Alfa Aesar Supplier No. H61431.22

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
5-Bromo-2-fluoro-4-methylpyridine is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

Solubility
Slightly soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 864830-16-0
Molecular Formula C6H5BrFN
Molecular Weight (g/mol) 190.015
MDL Number MFCD09839213
InChI Key PFODFMZCBQUYNG-UHFFFAOYSA-N
Synonym 5-bromo-2-fluoro-4-picoline, 5-bromo-2-fluoro-4-methyl-pyridine, 2-fluoro-4-methyl-5-bromopyridine, pubchem3547, acmc-209q9w, ksc495s8j
PubChem CID 23157062
IUPAC Name 5-bromo-2-fluoro-4-methylpyridine
SMILES CC1=CC(=NC=C1Br)F

Specifications

Color Colorless to Yellow
Refractive Index 1.53
Quantity 100 g
Solubility Information Slightly soluble in water.
Formula Weight 190.01
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material 5-Bromo-2-fluoro-4-methylpyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.