missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Bromo-2-fluoro-4-methylpyridine, 96%, Thermo Scientific™
Click to view available options
Quantity:
5 g
25 g
100 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
5-Bromo-2-fluoro-4-methylpyridine is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.
Solubility
Slightly soluble in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
5-Bromo-2-fluoro-4-methylpyridine is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.
Solubility
Slightly soluble in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications
Color | Colorless to Yellow |
Refractive Index | 1.53 |
Quantity | 25 g |
Solubility Information | Slightly soluble in water. |
Formula Weight | 190.01 |
Percent Purity | 96% |
Physical Form | Liquid |
Chemical Name or Material | 5-Bromo-2-fluoro-4-methylpyridine |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'