missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-ethoxypyridine, 97%, Thermo Scientific™

Product Code. 11332169
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11332169 1 g 1g
11342169 5 g 5g
11352169 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11332169 Supplier Thermo Scientific Alfa Aesar Supplier No. H27197.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 55849-30-4
Molecular Formula C7H8BrNO
Molecular Weight (g/mol) 202.051
MDL Number MFCD00234311
InChI Key WQXZKMUZWPUZGL-UHFFFAOYSA-N
Synonym 2-ethoxy-5-bromopyridine, 3-bromo-6-ethoxypyridine, 5-bromo-2-ethoxy-pyridine, pyridine, 5-bromo-2-ethoxy, zlchem 405, pubchem6235, 5-bromo-ethoxy-pyridine, acmc-1avox, 5-bromo-2-ethoxy pyridine, ksc497o6h
PubChem CID 817237
IUPAC Name 5-bromo-2-ethoxypyridine
SMILES CCOC1=NC=C(C=C1)Br

Specifications

Melting Point 32°C to 36°C
Flash Point >110°C (230°F)
Quantity 1 g
Formula Weight 202.05
Percent Purity 97%
Chemical Name or Material 5-Bromo-2-ethoxypyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.