missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-ethoxypyridine, 97%, Thermo Scientific™

Product Code. 11368181 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11368181 1 g 1g
11966431 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11368181 Supplier Thermo Scientific Acros Supplier No. 442310010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 55849-30-4
Molecular Formula C7H8BrNO
Molecular Weight (g/mol) 202.05
InChI Key WQXZKMUZWPUZGL-UHFFFAOYSA-N
Synonym 2-ethoxy-5-bromopyridine, 3-bromo-6-ethoxypyridine, 5-bromo-2-ethoxy-pyridine, pyridine, 5-bromo-2-ethoxy, zlchem 405, pubchem6235, 5-bromo-ethoxy-pyridine, acmc-1avox, 5-bromo-2-ethoxy pyridine, ksc497o6h
PubChem CID 817237
IUPAC Name 5-bromo-2-ethoxypyridine
SMILES CCOC1=NC=C(C=C1)Br

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 34°C to 37°C
Color White to Yellow
Boiling Point 107°C (33.0 Torr)
Flash Point 103°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 202.05
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 5-Bromo-2-ethoxypyridine
Show More Show Less
compliance-icons
Product Identifier
  • 5-Bromo-2-ethoxypyridine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.