missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Bromo-2'-deoxyuridine, Fisher BioReagents
Description
5-Bromo-2'-Deoxyuridine is a thymidine analog used as a mutagen in genetic research.
Specifications
Specifications
| Melting Point | 191°C |
| Color | White |
| Infrared Spectrum | Conforms to reference |
| Assay Percent Range | >99% |
| Packaging | Amber Glass |
| Quantity | 5 g |
| Merck Index | 15, 1461 |
| Formula Weight | 307.1 |
| Physical Form | Powder or Solid |
| Chemical Name or Material | 5-Bromo-2'-Deoxyuridine, Off-white Powder |
Safety and Handling
Product Identifier
- 5-Bromo-2'-deoxyuridine
- Warning
- Serious eye damage/eye irritation Category 2
- Germ cell mutagenicity Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H319-Causes serious eye irritation.
- H341-Suspected of causing genetic defects.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?