missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 5-Bromo-2'-deoxyuridine, 99+%

Product Code. p-3777604 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
500 mg
1 g
2.5 g
10 g
Packaging:
Glass bottle
Unit Size:
10g
1g
2.5g
500mg
This item is not returnable. View return policy

Product Code. 10733041

Brand: Thermo Scientific Chemicals 228595000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 59-14-3 | C9H11BrN2O5 | 307.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 59-14-3
Molecular Formula C9H11BrN2O5
Molecular Weight (g/mol) 307.10
MDL Number MFCD00006529
InChI Key WOVKYSAHUYNSMH-RRKCRQDMSA-N
Synonym 5-bromo-2'-deoxyuridine, broxuridine, bromodeoxyuridine, brdu, 5-bromodeoxyuridine, 5-brdu, budr, 5-bromouracil deoxyriboside, bromouracil deoxyriboside, 5-bromodesoxyuridine
PubChem CID 6035
ChEBI CHEBI:472552
IUPAC Name 5-bromo-1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidine-2,4-dione
SMILES OC[C@H]1O[C@H](C[C@@H]1O)N1C=C(Br)C(=O)NC1=O

Specifications

Absorbance 0.47 to 0.53 (A250/A260),1.87 to 1.93 (A280/A260),1.45 to 1.55 (A290/A260)
CAS Min % 99.0
CAS Max % 100.0
Melting Point 191.0°C to 194.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 99% min. (HPLC)
Packaging Glass bottle
Quantity 500 mg
Beilstein 24, IV, 1234
Merck Index 15, 1461
Specific Rotation + 31.00
Solubility Information Solubility in water: soluble. Other solubilities: soluble in ethanol,dmso (50mg/mL)
Residual Solvents 0.2% max. ethanol (GC), 0.1% max. methanol (GC)
Formula Weight 307.09
Percent Purity 99+%
Related Substances 1% max. (HPLC) Total impurities
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 5-Bromo-2′-deoxyuridine
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 5-Bromo-2'-deoxyuridine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Germ cell mutagenicity Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H341-Suspected of causing genetic defects.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.