Learn More
Lonza™ 5-Bromo-2'Deoxyuridine
Click to view available options
Quantity:
15 mg
Unit Size:
Each
Description
- Synthetic nucleoside that is an analog of thymidine.
- Commonly used in the detection of proliferating cells in living tissues.
- Routinely and extensively used to measure DNA synthesis and to label dividing cells.
Specifications
Specifications
| Product Type | Nucleosides |
| Quantity | 15 mg |