missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2'-deoxycytidine, MP Biomedicals™

Product Code. 11308489 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11308489 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11308489 Supplier MP Biomedicals Supplier No. 0210016690

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

5-Bromo-2'-deoxycytidine can be incorporated into DNA in the presence of DNA polymerase, replacing dCTP

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1022-79-3
Molecular Formula C9H12BrN3O4
Molecular Weight (g/mol) 306.116
InChI Key KISUPFXQEHWGAR-RRKCRQDMSA-N
Synonym 5-bromo-2'-deoxycytidine, 5-bromodeoxycytidine, 4-amino-5-bromo-1-2r,4s,5r-4-hydroxy-5-hydroxymethyl tetrahydrofuran-2-yl pyrimidin-2 1h-one, unii-6m3tp1aibh, brcdr, bromodeoxycytidine, 6m3tp1aibh, bcdr, 5-bromo-2'-deoxycytidine monohydrate, cytidine, 5-bromo-2'-deoxy
PubChem CID 472175
IUPAC Name 4-amino-5-bromo-1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidin-2-one
SMILES C1C(C(OC1N2C=C(C(=NC2=O)N)Br)CO)O

Specifications

Color White
Quantity 500 mg
Formula Weight 306.1 (Anhydrous)
Physical Form Powder
Chemical Name or Material 5-Bromo-2'-deoxycytidine monohydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.