missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-cyanopyrimidine, 95%

Product Code. 15467888
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15467888 5 g 5g
15457888 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15467888 Supplier Thermo Scientific Alfa Aesar Supplier No. H61363.06
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 38275-57-9 | C5H2BrN3 | 183.996 g/mol

5-Bromo-2-cyanopyrimidine is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Bromo-2-cyanopyrimidine is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

Solubility
Slightly soluble in water.

Notes
Moisture Sensitive. Store away form water/moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 38275-57-9
Molecular Formula C5H2BrN3
Molecular Weight (g/mol) 183.996
MDL Number MFCD02940446
InChI Key VPQICCOHFSGBMA-UHFFFAOYSA-N
Synonym 5-bromo-2-cyanopyrimidine, 5-bromo-2-pyrimidinecarbonitrile, 2-pyrimidinecarbonitrile, 5-bromo, 5-bromo-pyrimidine-2-carbonitile, 2-cyano-5-bromopyrimidine, pubchem5278, acmc-1ad2b, 5-bromo-2-cyano-pyrimidine, ksc497m1f, 5-bromo-pyrimidine-2-carbonitrile
PubChem CID 686546
IUPAC Name 5-bromopyrimidine-2-carbonitrile
SMILES C1=C(C=NC(=N1)C#N)Br

Specifications

Color White to Yellow
Quantity 5 g
UN Number UN3439
Beilstein 775324
Sensitivity Moisture sensitive
Solubility Information Slightly soluble in water.
Formula Weight 183.99
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 5-Bromo-2-cyanopyrimidine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.