missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-chlorotoluene, 98%, Thermo Scientific™

Product Code. 11350027 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
11350027 25 g Glass bottle 25g
11340027 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11350027 Supplier Thermo Scientific Acros Supplier No. 352410250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 54932-72-8
Molecular Formula C7H6BrCl
Molecular Weight (g/mol) 205.48
InChI Key OZFQMHJKAODEON-UHFFFAOYSA-N
Synonym 5-bromo-2-chlorotoluene, benzene, 4-bromo-1-chloro-2-methyl, 2-chloro-5-bromotoluene, pubchem3604, 5-bromo-2-chlorotoulene, acmc-209lk8, ksc493o2d, ozfqmhjkaodeon-uhfffaoysa, attercop-chm at115283
PubChem CID 609899
IUPAC Name 4-bromo-1-chloro-2-methylbenzene
SMILES CC1=C(C=CC(=C1)Br)Cl

Specifications

CAS Min % 97.5
CAS Max % 100.0
Density 1.5490g/mL
Quantity 25 g
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Liquid
Refractive Index 1.5735 to 1.5755
Assay Percent Range 97.5% min. (GC)
Specific Gravity 1.549
Boiling Point 90.0°C to 93.0°C (12.0 mmHg)
Formula Weight 205.48
Percent Purity 98%
Chemical Name or Material 5-Bromo-2-chlorotoluene
Show More Show Less
compliance-icons
Product Identifier
  • 5-Bromo-2-chlorotoluene
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.