missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-chloronicotinic acid, 96%

Product Code. 15477658
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15477658 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15477658 Supplier Thermo Scientific Alfa Aesar Supplier No. H61154.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 29241-65-4 | C6H3BrClNO2 | 236.45 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 29241-65-4
Molecular Formula C6H3BrClNO2
MDL Number MFCD03844847
Quantity 100 g
Synonym 5-bromo-2-chloronicotinic acid, 5-bromo-2-chloro-3-pyridinecarboxylic acid, 3-pyridinecarboxylic acid, 5-bromo-2-chloro, 5-bromo-2-chloro-nicotinic acid, 5-bromo-3-carboxy-2-chloropyridine, 2-chloro-5-bromonicotinic acid, 5-bromo-2-chloronicotinicacid, pubchem12967, 5-bromo-2-chloro-pyridine-3-carboxylic acid, acmc-209h7p
InChI Key UKNYSJCAGUXDOQ-UHFFFAOYSA-N
SMILES OC(=O)C1=CC(Br)=CN=C1Cl
IUPAC Name 5-bromo-2-chloropyridine-3-carboxylic acid
Molecular Weight (g/mol) 236.45
PubChem CID 2763342
Formula Weight 236.45
Percent Purity 96%
Physical Form Solid
Chemical Name or Material 5-Bromo-2-chloronicotinic acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.