missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-chlorobenzotrifluoride, 98+%, Thermo Scientific™

Product Code. 11458713
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
250 g
Unit Size:
10g
250g
25g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11458713 10 g 10g
11354669 5 g 5g
11364669 25 g 25g
11468713 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11458713 Supplier Thermo Scientific Alfa Aesar Supplier No. A10477.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 445-01-2
Molecular Formula C7H3BrClF3
Molecular Weight (g/mol) 259.45
MDL Number MFCD00000601
InChI Key XGOCKBMEZPNDPJ-UHFFFAOYSA-N
Synonym 5-bromo-2-chlorobenzotrifluoride, 4-bromo-1-chloro-2-trifluoromethyl benzene, 2-chloro-5-bromobenzotrifluoride, 5-bromo-2-chloro-benzenetrifluoride, 5-bromo-2-chlorotrifluoride, 3-bromo-6-chlorobenzotrifluoride, benzene, 4-bromo-1-chloro-2-trifluoromethyl, 5-bromo-2-chloro-alpha,alpha,alpha-trifluorotoluene, 2-chloro-5-bromo-trifluorotoluene, 4-bromo-1-chloro-2-trifluoromethyl-benzene
PubChem CID 67959
IUPAC Name 4-bromo-1-chloro-2-(trifluoromethyl)benzene
SMILES FC(F)(F)C1=CC(Br)=CC=C1Cl

Specifications

Melting Point -21°C
Density 1.745
Boiling Point 195°C
Flash Point 81°C (177°F)
Refractive Index 1.507
Quantity 10 g
Beilstein 2098752
Formula Weight 259.44
Percent Purity ≥98%
Chemical Name or Material 5-Bromo-2-chlorobenzotrifluoride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.