missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-chlorobenzoic acid, 98+%

Product Code. 11434893
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11434893 10 g 10g
11444893 50 g 50g
11454893 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11434893 Supplier Thermo Scientific Alfa Aesar Supplier No. A12218.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 21739-92-4 | C7H4BrClO2 | 235.461 g/mol

5-Bromo-2-chlorobenzoic acid was used to evaluate the homogeneity of organic powder. It can be used to produce 2-chloro-5-deuteriobenzoic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Bromo-2-chlorobenzoic acid was used to evaluate the homogeneity of organic powder. It can be used to produce 2-chloro-5-deuteriobenzoic acid.

Solubility
Soluble in water 2.63 g/L @20°C.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21739-92-4
Molecular Formula C7H4BrClO2
Molecular Weight (g/mol) 235.461
MDL Number MFCD00002415
InChI Key FGERXQWKKIVFQG-UHFFFAOYSA-N
Synonym 2-chloro-5-bromobenzoic acid, benzoic acid, 5-bromo-2-chloro, 5-bromo-2-chlorobenzoicacid, 5-bromo-2-chloro benzoic acid, 5-bromo-2-chloro-benzoic acid, bromo 5--2-chlorobenzoic acid, pubchem3587, acmc-209fnm, 5-bromo-2-chorobenzoic acid, ksc204k3r
PubChem CID 33127
IUPAC Name 5-bromo-2-chlorobenzoic acid
SMILES C1=CC(=C(C=C1Br)C(=O)O)Cl

Specifications

Melting Point 157°C to 162°C
Odor Odorless
Quantity 10 g
Beilstein 2691432
Solubility Information Soluble in water 2.63g/L @20°C.
Formula Weight 235.47
Percent Purity ≥98%
Chemical Name or Material 5-Bromo-2-chlorobenzoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.